siRNA Details
| Gene Symbol: RAB38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022337.2 | NP_071732.1 | 3 | 760 | ||
| GenBank mRNA | AF136172.1 | - | 3 | 688 | ||
| AF235022.1 | - | 3 | 724 | |||
| AL546159.3 | - | 3 | 759 | |||
| BC015808.1 | - | 3 | 726 | 3916084 | ||

RAB38
Genomic Map