siRNA Details
| Gene Symbol: RNF133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139175.1 | NP_631914.1 | 1 | 1036 | ||
| GenBank mRNA | AF447589.1 | - | 1 | 1036 | ||
| AK098524.1 | - | 1 | 1038 | |||
| BC022038.2 | - | 1 | 1015 | 4823227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF133| Gene Symbol: RNF133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139175.1 | NP_631914.1 | 1 | 1036 | ||
| GenBank mRNA | AF447589.1 | - | 1 | 1036 | ||
| AK098524.1 | - | 1 | 1038 | |||
| BC022038.2 | - | 1 | 1015 | 4823227 | ||