siRNA Details
| Gene Symbol: NDUFA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318195.1 | NP_001305124.1 | 2 | 304 | ||
| NM_014222.2 | NP_055037.1 | 2 | 304 | |||
| GenBank mRNA | AF044953.1 | - | 2 | 229 | ||
| AK301520.1 | - | 2 | 272 | |||
| AK314135.1 | - | 2 | 264 | |||
| BC001016.2 | - | 2 | 208 | 3345138 | ||
| BU602971.1 | - | 2 | 306 | |||

NDUFA8
Genomic Map