siRNA Details
| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136635.1 | - | 6 | 900 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB1B| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136635.1 | - | 6 | 900 | ||