siRNA Details
| Gene Symbol: TUBB4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006088.5 | NP_006079.1 | 4 | 683 | ||
| GenBank mRNA | AK000560.1 | - | 4 | 655 | ||
| AK021414.1 | - | 4 | 511 | |||
| AK026167.1 | - | 4 | 654 | |||
| AK026594.1 | - | 4 | 655 | |||
| AK026609.1 | - | 4 | 655 | |||
| AK098686.1 | - | 4 | 584 | |||
| AY167990.1 | - | 4 | 581 | |||
| BC001911.1 | - | 4 | 618 | 3544236 | ||
| BC002783.1 | - | 4 | 628 | 3614023 | ||
| BC002885.2 | - | 4 | 625 | 3944735 | ||
| BC004188.2 | - | 4 | 630 | 2964559 | ||
| BC007889.2 | - | 4 | 619 | 4127195 | ||
| BC012835.2 | - | 4 | 630 | 3603755 | ||
| BC019359.1 | - | 4 | 621 | 4024979 | ||
| BC019829.1 | - | 4 | 644 | 5110038 | ||
| BC024038.1 | - | 4 | 619 | 3616531 | ||
| BC029529.1 | - | 4 | 655 | 5274343 | ||
| BC039175.1 | - | 4 | 628 | 4385885 | ||
| BC071889.1 | - | 4 | 619 | 4905049 | ||
| BX648521.1 | - | 3 | 1129 | |||

TUBB4B
Genomic Map