siRNA Details
| Gene Symbol: RRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006270.4 | NP_006261.1 | 6 | 823 | ||
| GenBank mRNA | BC016286.1 | - | 6 | 802 | 3910343 | |
| BC016318.1 | - | 6 | 827 | 4073829 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRAS| Gene Symbol: RRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006270.4 | NP_006261.1 | 6 | 823 | ||
| GenBank mRNA | BC016286.1 | - | 6 | 802 | 3910343 | |
| BC016318.1 | - | 6 | 827 | 4073829 | ||