siRNA Details
Gene Symbol: WTH3DI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077637.1 | NP_001071105.1 | 1 | 2031 |
Gene Symbol: RAB6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032144.2 | NP_115520.2 | 1 | 2046 | ||
GenBank mRNA | AK055504.1 | - | 2 | 1433 | ||
AL136727.1 | - | 1 | 2047 |
