siRNA Details
| Gene Symbol: TUBA3FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC009991.2 | - | 2, 3 | 572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBA3FP| Gene Symbol: TUBA3FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC009991.2 | - | 2, 3 | 572 | ||