siRNA Details
| Gene Symbol: TUBA3FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003608.1 | 3 | 844 | |||
| GenBank mRNA | AK057257.1 | - | 3 | 796 | ||
| AK315686.1 | - | 3 | 792 | |||
| BC009991.2 | - | 3 | 698 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUBA3FP| Gene Symbol: TUBA3FP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003608.1 | 3 | 844 | |||
| GenBank mRNA | AK057257.1 | - | 3 | 796 | ||
| AK315686.1 | - | 3 | 792 | |||
| BC009991.2 | - | 3 | 698 | |||