siRNA Details
| Gene Symbol: GPR150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199243.1 | NP_954713.1 | 1 | 250 | ||
| GenBank mRNA | BC030197.1 | - | 1 | 448 | ||
| JF810884.1 | - | 1 | 250 | |||
More Information
Other Species Targeted
Set Membership: |
|

GPR150
Genomic Map