siRNA Details
| Gene Symbol: USP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017029164.1 | XP_016884653.1 | 6 | 520 | ||
| GenBank mRNA | AJ583822.1 | - | 5 | 493 | ||
| AJ586979.1 | - | 4 | 430 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP41| Gene Symbol: USP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017029164.1 | XP_016884653.1 | 6 | 520 | ||
| GenBank mRNA | AJ583822.1 | - | 5 | 493 | ||
| AJ586979.1 | - | 4 | 430 | |||