siRNA Details
| Gene Symbol: SRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321053.1 | NP_001307982.1 | 4 | 992 | ||
| NM_001321054.1 | NP_001307983.1 | 2 | 629 | |||
| NM_002727.3 | NP_002718.2 | 3 | 777 | |||
| XM_017016392.1 | XP_016871881.1 | 4 | 1405 | |||
| GenBank mRNA | BC015516.1 | - | 3 | 687 | 3883161 | |
| BQ051861.1 | - | 2 | 540 | |||
| CD677686.1 | - | 1 | 363 | |||
| X17042.1 | - | 3 | 671 | |||

SRGN
Genomic Map