siRNA Details
| Gene Symbol: OSBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002556.2 | NP_002547.1 | 1 | 784 | ||
| GenBank mRNA | AF185696.1 | - | 1 | 784 | ||
| BC011581.1 | - | 1 | 372 | 4560111 | ||
| M86917.1 | - | 1 | 784 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OSBP| Gene Symbol: OSBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002556.2 | NP_002547.1 | 1 | 784 | ||
| GenBank mRNA | AF185696.1 | - | 1 | 784 | ||
| BC011581.1 | - | 1 | 372 | 4560111 | ||
| M86917.1 | - | 1 | 784 | |||