siRNA Details
| Gene Symbol: PRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002761.2 | NP_002752.1 | 2 | 376 | ||
| GenBank mRNA | AY651260.1 | - | 2 | 357 | ||
| BC003673.1 | - | 2 | 369 | 3935638 | ||
| CD360017.1 | - | 2 | 350 | |||
| Y00443.1 | - | 2 | 379 | |||

PRM1
Genomic Map