siRNA Details
| Gene Symbol: SNAPIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012437.5 | NP_036569.1 | 4 | 462 | ||
| NR_052019.1 | 3 | 415 | ||||
| NR_052020.1 | 3 | 401 | ||||
| GenBank mRNA | AF086837.1 | - | 4 | 372 | ||
| AK024555.1 | - | 4 | 439 | |||
| BC000761.2 | - | 4 | 391 | 2821705 | ||
| BC004494.2 | - | 4 | 392 | 3830132 | ||
| BE278772.1 | - | 3 | 316 | |||
| BI117540.1 | - | 463 | ||||
| BM051828.1 | - | 4 | 448 | |||
| BT006753.1 | - | 4 | 372 | |||
| CD671119.1 | - | 3 | 350 | |||

SNAPIN
Genomic Map