siRNA Details
| Gene Symbol: BANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083907.2 | NP_001077376.2 | 3 | 632 | ||
| NM_001127507.2 | NP_001120979.2 | 2 | 464 | |||
| NM_017935.4 | NP_060405.4 | 3 | 863 | |||
| XM_017008337.1 | XP_016863826.1 | 3 | 610 | |||
| GenBank mRNA | AB063170.1 | - | 2 | 499 | ||
| AK000713.1 | - | 3 | 579 | |||
| AK091523.1 | - | 7 | 964 | |||
| AK292133.1 | - | 3 | 707 | |||
| AL834291.1 | - | 3 | 682 | |||
| BC032241.1 | - | 3 | 731 | 5214147 | ||
| EU051376.1 | - | 2 | 202 | |||

BANK1
Genomic Map