siRNA Details
| Gene Symbol: OSBPL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022776.4 | NP_073613.2 | 9 | 2087 | ||
| GenBank mRNA | AF392454.1 | - | 9 | 1658 | ||
| AK023074.1 | - | 4 | 484 | |||
| AK023226.1 | - | 4 | 517 | |||
| AK292702.1 | - | 9 | 1439 | |||
| BC010422.1 | - | 4 | 671 | |||
| BC065213.1 | - | 9 | 1437 | 3916115 | ||

OSBPL11
Genomic Map