siRNA Details
| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242508.1 | NP_001229437.1 | 7 | 724 | ||
| NM_018030.4 | NP_060500.3 | 5 | 1093 | |||
| NM_080597.3 | NP_542164.2 | 19 | 1980 | |||
| XM_006722380.3 | XP_006722443.1 | 18 | 1893 | |||
| XM_017025530.1 | XP_016881019.1 | 19 | 2272 | |||
| XM_017025531.1 | XP_016881020.1 | 18 | 1765 | |||
| XM_017025533.1 | XP_016881022.1 | 5 | 576 | |||
| XM_017025534.1 | XP_016881023.1 | 5 | 3104 | |||
| GenBank mRNA | AF274714.1 | - | 5 | 1093 | ||
| AF323726.2 | - | 19 | 1937 | |||
| AF392449.1 | - | 5 | 714 | |||
| AK001079.1 | - | 6 | 621 | |||
| AK096358.1 | - | 19 | 1980 | |||
| AK292895.1 | - | 20 | 1912 | |||
| AK301862.1 | - | 7 | 724 | |||
| AK314552.1 | - | 5 | 544 | |||
| BC063420.1 | - | 5 | 779 | |||
| BC073854.1 | - | 3 | 300 | |||
| BC098384.1 | - | 5 | 772 | |||
| BC136452.1 | - | 19 | 1877 | 9020441 | ||
| BC144117.1 | - | 19 | 1877 | 9052637 | ||

OSBPL1A
Genomic Map