siRNA Details
| Gene Symbol: CES5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143685.1 | NP_001137157.1 | 12, 13 | 1650 | ||
| NM_001190158.1 | NP_001177087.1 | 13, 14 | 1734 | |||
| NM_145024.2 | NP_659461.1 | 11, 12 | 1500 | |||
| GenBank mRNA | AK056109.1 | - | 11, 12 | 1500 | ||
| AK090997.1 | - | 12, 13 | 1576 | |||
| AK294334.1 | - | 13, 14 | 1734 | |||
| AY907669.1 | - | 12, 13 | 1577 | |||
| BC039073.1 | - | 12, 13 | 1663 | |||
| BC069501.1 | - | 12, 13 | 1621 | 7262427 | ||
| BC069548.1 | - | 11, 12 | 1470 | 7262439 | ||
| BC117126.1 | - | 12, 13 | 1623 | 40125677 | ||
| BC143692.1 | - | 12, 13 | 1623 | 9052206 | ||
| EU794678.1 | - | 11, 12 | 1500 | |||

CES5A
Genomic Map