siRNA Details
| Gene Symbol: ITGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181501.1 | NP_852478.1 | 7 | 711 | ||
| GenBank mRNA | AK300423.1 | - | 7 | 876 | ||
| BC137121.1 | - | 7 | 710 | 9021118 | ||
| BC137122.1 | - | 7 | 710 | 9021119 | ||
| X68742.1 | - | 6 | 575 | |||

ITGA1
Genomic Map