siRNA Details
| Gene Symbol: OPLAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017570.4 | NP_060040.1 | 13, 14 | 1954 | ||
| XM_011516960.1 | XP_011515262.1 | 13, 14 | 2230 | |||
| GenBank mRNA | AB122018.1 | - | 12, 13 | 1850 | ||
| AL096750.1 | - | 5, 6 | 939 | |||
| BC036944.1 | - | 8, 9 | 1141 | |||
| BC142672.1 | - | 13, 14 | 1954 | 40148181 | ||
| BC150206.1 | - | 13, 14 | 1932 | 40148883 | ||

OPLAH
Genomic Map