siRNA Details
| Gene Symbol: GRINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000837.1 | NP_000828.1 | 3, 4 | 778 | ||
| NM_001009184.1 | NP_001009184.1 | 3, 4 | 666 | |||
| GenBank mRNA | AB097026.1 | - | 3, 4 | 590 | ||
| AK127640.1 | - | 3, 4 | 814 | |||
| AK294127.1 | - | 4, 5 | 523 | |||
| BC041788.1 | - | 3, 4 | 624 | 4738372 | ||
| BC065834.1 | - | 2, 3 | 532 | |||
| BC084553.1 | - | 2, 3 | 515 | 6206624 | ||

GRINA
Genomic Map