siRNA Details
| Gene Symbol: KNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000893.3 | NP_000884.1 | 1 | 386 | ||
| NM_001102416.2 | NP_001095886.1 | 1 | 386 | |||
| NM_001166451.1 | NP_001159923.1 | 1 | 386 | |||
| GenBank mRNA | AI133186.1 | - | 1 | 296 | ||
| AK223589.1 | - | 1 | 335 | |||
| AK290839.1 | - | 1 | 285 | |||
| AK298440.1 | - | 2 | 300 | |||
| AK303768.1 | - | 1 | 366 | |||
| AK315230.1 | - | 1 | 338 | |||
| BC026253.1 | - | 1 | 155 | |||
| BC060039.1 | - | 1 | 289 | 4593077 | ||
| BG430928.1 | - | 1 | 365 | |||
| CD014130.1 | - | 1 | 348 | |||
| DA537547.1 | - | 1 | 364 | |||
| DC362362.1 | - | 1 | 386 | |||
| K02566.1 | - | 1 | 201 | |||

KNG1
Genomic Map