siRNA Details
| Gene Symbol: PSMB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130725.1 | NP_001124197.1 | 3 | 890 | ||
| NM_001144932.2 | NP_001138404.1 | 4 | 1328 | |||
| NM_002797.4 | NP_002788.1 | 3 | 1243 | |||
| GenBank mRNA | AK225888.1 | - | 3 | 1008 | ||
| AK300714.1 | - | 4 | 1073 | |||
| BC057840.1 | - | 3 | 998 | 4795732 | ||
| BC107720.1 | - | 3 | 994 | 6738409 | ||
| BX538001.1 | - | 3 | 990 | |||

PSMB5
Genomic Map