siRNA Details
| Gene Symbol: PSMB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270481.1 | NP_001257410.1 | 5 | 597 | ||
| NM_002798.2 | NP_002789.1 | 5 | 597 | |||
| GenBank mRNA | AK311768.1 | - | 5 | 560 | ||
| BC000835.2 | - | 5 | 564 | 3456009 | ||
| BM553399.1 | - | 5 | 544 | |||
| BQ691763.1 | - | 5 | 579 | |||
| BT009809.1 | - | 5 | 546 | |||
| BU848746.1 | - | 5 | 529 | |||
| CR457065.1 | - | 5 | 546 | |||
| D29012.1 | - | 5 | 555 | |||
| X61971.1 | - | 3 | 369 | |||

PSMB6
Genomic Map