siRNA Details
| Gene Symbol: CENPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286734.1 | NP_001273663.1 | 15, 16 | 1565 | ||
| NM_001813.2 | NP_001804.2 | 15, 16 | 1565 | |||
| XM_011531544.2 | XP_011529846.1 | 15, 16 | 1558 | |||
| XM_011531545.2 | XP_011529847.1 | 15, 16 | 1564 | |||
| XM_011531546.2 | XP_011529848.1 | 15, 16 | 1564 | |||
| XM_011531547.2 | XP_011529849.1 | 15, 16 | 1564 | |||
| XM_011531548.2 | XP_011529850.1 | 15, 16 | 1564 | |||
| XM_011531549.2 | XP_011529851.1 | 15, 16 | 1564 | |||
| XM_017007659.1 | XP_016863148.1 | 15, 16 | 1564 | |||
| GenBank mRNA | AB209996.1 | - | 15, 16 | 1490 | ||
| AK290362.1 | - | 15, 16 | 1566 | |||
| Z15005.1 | - | 15, 16 | 1565 | |||

CENPE
Genomic Map