siRNA Details
| Gene Symbol: SFRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003014.3 | NP_003005.2 | 6 | 2313 | ||
| GenBank mRNA | AF026692.1 | - | 6 | 2182 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFRP4| Gene Symbol: SFRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003014.3 | NP_003005.2 | 6 | 2313 | ||
| GenBank mRNA | AF026692.1 | - | 6 | 2182 | ||
Set Membership: |
|