siRNA Details
| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075140.1 | - | 6 | 548 | ||
| AK309540.1 | - | 7 | 577 | |||
| AY358081.1 | - | 6 | 537 | |||
| AY358283.1 | - | 6 | 574 | |||
| BC020731.1 | - | 6 | 548 | 4692272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FCN3| Gene Symbol: FCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK075140.1 | - | 6 | 548 | ||
| AK309540.1 | - | 7 | 577 | |||
| AY358081.1 | - | 6 | 537 | |||
| AY358283.1 | - | 6 | 574 | |||
| BC020731.1 | - | 6 | 548 | 4692272 | ||