siRNA Details
| Gene Symbol: DDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257191.2 | NP_001244120.1 | 6 | 970 | ||
| NM_004397.5 | NP_004388.2 | 6 | 1042 | |||
| XM_005271417.3 | XP_005271474.1 | 6 | 987 | |||
| XM_011542644.1 | XP_011540946.1 | 6 | 1042 | |||
| XM_017017251.1 | XP_016872740.1 | 6 | 987 | |||
| GenBank mRNA | AK313244.1 | - | 6 | 970 | ||
| BC065007.1 | - | 5 | 901 | 6163439 | ||
| D17532.1 | - | 6 | 915 | |||
| Z11685.1 | - | 6 | 942 | |||

DDX6
Genomic Map