siRNA Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005065.5 | NP_005056.3 | 12 | 1314 | ||
| XM_005267988.2 | XP_005268045.1 | 12 | 1197 | |||
| GenBank mRNA | AB020335.1 | - | 12 | 1242 | ||
| AF052059.1 | - | 12 | 1290 | |||
| AK075511.1 | - | 12 | 1293 | |||

SEL1L
Genomic Map