siRNA Details
| Gene Symbol: CBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005188.3 | NP_005179.2 | 2 | 394 | ||
| XM_011543057.2 | XP_011541359.1 | 2 | 394 | |||
| GenBank mRNA | BC132733.1 | - | 2 | 323 | 40146755 | |
| BC136463.1 | - | 2 | 323 | 9020452 | ||
| BG722940.1 | - | 2 | 384 | |||
| DB457271.2 | - | 2 | 394 | |||
| X57110.1 | - | 2 | 400 | |||

CBL
Genomic Map