siRNA Details
| Gene Symbol: WDR46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005452.5 | NP_005443.3 | 2 | 581 | ||
| XM_017011484.1 | XP_016866973.1 | 2 | 295 | |||
| GenBank mRNA | AF447870.1 | - | 2 | 273 | ||
| AK292171.1 | - | 2 | 564 | |||
| AK292616.1 | - | 2 | 364 | |||
| AK297925.1 | - | 2 | 176 | |||
| AK315616.1 | - | 2 | 293 | |||
| BC000388.2 | - | 2 | 268 | 2821198 | ||
| BX439421.2 | - | 2 | 268 | |||
| DB272479.1 | - | 2 | 292 | |||

WDR46
Genomic Map