siRNA Details
| Gene Symbol: NECTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042724.1 | NP_001036189.1 | 2 | 494 | ||
| NM_002856.2 | NP_002847.1 | 2 | 494 | |||
| XM_011527192.2 | XP_011525494.1 | 2 | 494 | |||
| GenBank mRNA | AF058448.1 | - | 2 | 253 | ||
| AK291330.1 | - | 2 | 320 | |||
| BC003091.1 | - | 2 | 302 | 3503222 | ||
| CN410857.1 | - | 2 | 466 | |||
| CR456818.1 | - | 2 | 104 | |||
| X80038.1 | - | 2 | 104 | |||

NECTIN2
Genomic Map