siRNA Details
| Gene Symbol: ARPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024959.2 | NP_001020130.1 | 6 | 1124 | ||
| NM_001024960.2 | NP_001020131.1 | 6 | 979 | |||
| NM_001198780.1 | NP_001185709.1 | 6 | 978 | |||
| NM_005718.4 | NP_005709.1 | 6 | 1490 | |||
| GenBank mRNA | AK074205.1 | - | 1 | 1593 | ||
| BC012596.2 | - | 5 | 909 | |||

ARPC4
Genomic Map