siRNA Details
| Gene Symbol: TGOLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206840.1 | NP_001193769.1 | 2 | 707 | ||
| NM_001206841.1 | NP_001193770.1 | 2 | 707 | |||
| NM_001206844.1 | NP_001193773.1 | 2 | 707 | |||
| NM_006464.3 | NP_006455.2 | 2 | 707 | |||
| GenBank mRNA | AF027515.1 | - | 2 | 435 | ||
| AF027516.1 | - | 2 | 435 | |||
| AK222829.1 | - | 2 | 399 | |||
| AK223063.1 | - | 2 | 398 | |||
| AK312479.1 | - | 2 | 400 | |||
| BC008461.1 | - | 2 | 400 | 4272702 | ||
| BC028219.1 | - | 2 | 396 | 5217823 | ||
| BX640868.1 | - | 2 | 663 | |||
| U62390.1 | - | 2 | 435 | |||
| X94333.1 | - | 2 | 391 | |||

TGOLN2
Genomic Map