siRNA Details
| Gene Symbol: TGOLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206840.1 | NP_001193769.1 | 4 | 3024 | ||
| NM_001206841.1 | NP_001193770.1 | 4 | 2983 | |||
| NM_001206844.1 | NP_001193773.1 | 5 | 2792 | |||
| NM_006464.3 | NP_006455.2 | 4 | 2966 | |||
| GenBank mRNA | AK126465.1 | - | 6 | 2189 | ||
| AW630597.1 | - | 1 | 414 | |||
| BX640733.1 | - | 3 | 1867 | |||
| BX640868.1 | - | 5 | 2748 | |||

TGOLN2
Genomic Map