siRNA Details
| Gene Symbol: DDX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007204.4 | NP_009135.4 | 4 | 941 | ||
| GenBank mRNA | AF106019.1 | - | 4 | 594 | ||
| AF155803.1 | - | 4 | 589 | |||
| AF171063.1 | - | 4 | 877 | |||
| AK001506.1 | - | 4 | 603 | |||
| AK301697.1 | - | 3 | 442 | |||
| AK314312.1 | - | 4 | 586 | |||
| AL133598.1 | - | 1 | 96 | |||
| BC011556.1 | - | 4 | 594 | 4651697 | ||
| BC031062.1 | - | 4 | 603 | 5273709 | ||
| BC034953.2 | - | 4 | 912 | 4824324 | ||

DDX20
Genomic Map