siRNA Details
| Gene Symbol: CNTNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014141.5 | NP_054860.1 | 10 | 2033 | ||
| XM_017011950.1 | XP_016867439.1 | 10 | 2359 | |||
| GenBank mRNA | AB020675.1 | - | 10 | 1542 | ||
| AF319045.1 | - | 10 | 1657 | |||
| AK315113.1 | - | 10 | 1622 | |||
| BC093780.1 | - | 10 | 1560 | 7939625 | ||
| BC113373.1 | - | 10 | 1560 | 8322425 | ||
| KJ534811.1 | - | 9 | 1406 | |||

CNTNAP2
Genomic Map