siRNA Details
| Gene Symbol: HSPB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057295.1 | - | 3 | 1687 | ||
| AK075151.1 | - | 2 | 1500 | |||
| AK092275.1 | - | 4 | 1212 | |||
| AK095911.1 | - | 3 | 942 | |||
| BC006319.2 | - | 3 | 1355 | 4131506 | ||
| BX537990.1 | - | 3 | 926 | |||
| CR749298.1 | - | 3 | 954 | |||
| CR749419.1 | - | 3 | 954 | |||

HSPB7
Genomic Map