siRNA Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166252.1 | NP_001159724.1 | 2 | 513 | ||
| NM_014839.4 | NP_055654.2 | 2 | 513 | |||
| XM_011542498.2 | XP_011540800.1 | 2 | 461 | |||
| GenBank mRNA | AB007924.3 | - | 2 | 509 | ||
| AF357013.1 | - | 2 | 286 | |||
| AF541281.1 | - | 2 | 520 | |||
| AK291369.1 | - | 2 | 313 | |||
| AK300361.1 | - | 2 | 257 | |||
| AL834390.1 | - | 1 | 26 | |||
| AY304518.1 | - | 2 | 461 | |||
| BC042963.1 | - | 2 | 331 | |||

PLPPR4
Genomic Map