siRNA Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166252.1 | NP_001159724.1 | 2 | 520 | ||
| NM_014839.4 | NP_055654.2 | 2 | 520 | |||
| XM_011542498.2 | XP_011540800.1 | 2 | 468 | |||
| GenBank mRNA | AB007924.3 | - | 2 | 516 | ||
| AF357013.1 | - | 2 | 293 | |||
| AF541281.1 | - | 2 | 527 | |||
| AK291369.1 | - | 2 | 320 | |||
| AK300361.1 | - | 2 | 264 | |||
| AL834390.1 | - | 1 | 33 | |||
| AY304518.1 | - | 2 | 468 | |||
| BC042963.1 | - | 2 | 338 | |||

PLPPR4
Genomic Map