siRNA Details
| Gene Symbol: SZT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015284.3 | NP_056099.3 | 39 | 5696 | ||
| XM_005270686.3 | XP_005270743.1 | 41 | 5928 | |||
| XM_006710501.3 | XP_006710564.1 | 40 | 5815 | |||
| XM_011541106.2 | XP_011539408.1 | 40 | 5966 | |||
| XM_011541107.2 | XP_011539409.1 | 37 | 5463 | |||
| XM_017000819.1 | XP_016856308.1 | 41 | 5775 | |||
| XM_017000820.1 | XP_016856309.1 | 40 | 5757 | |||
| XM_017000821.1 | XP_016856310.1 | 37 | 5449 | |||
| GenBank mRNA | AB007936.2 | - | 25 | 3548 | ||
| BC151232.1 | - | 25 | 3548 | 8860124 | ||

SZT2
Genomic Map