siRNA Details
| Gene Symbol: SZT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015284.3 | NP_056099.3 | 51 | 7145 | ||
| XM_005270686.3 | XP_005270743.1 | 53 | 7377 | |||
| XM_006710501.3 | XP_006710564.1 | 52 | 7264 | |||
| XM_011541106.2 | XP_011539408.1 | 52 | 7415 | |||
| XM_011541107.2 | XP_011539409.1 | 49 | 6912 | |||
| XM_017000819.1 | XP_016856308.1 | 53 | 7224 | |||
| XM_017000820.1 | XP_016856309.1 | 52 | 7206 | |||
| XM_017000821.1 | XP_016856310.1 | 49 | 6898 | |||
| GenBank mRNA | AB007936.2 | - | 37 | 4997 | ||
| BC151232.1 | - | 37 | 4997 | 8860124 | ||

SZT2
Genomic Map