siRNA Details
| Gene Symbol: TM7SF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016551.2 | NP_057635.1 | 2 | 378 | ||
| XM_005253391.3 | XP_005253448.1 | 2 | 399 | |||
| XM_011520691.1 | XP_011518993.1 | 2 | 399 | |||
| XM_017019463.1 | XP_016874952.1 | 2 | 399 | |||
| GenBank mRNA | AB032470.1 | - | 2 | 177 | ||
| AK002031.1 | - | 2 | 144 | |||
| AK023085.1 | - | 2 | 366 | |||
| BC005176.2 | - | 2 | 303 | 3505386 | ||
| DA719107.1 | - | 2 | 384 | |||

TM7SF3
Genomic Map