siRNA Details
| Gene Symbol: TM7SF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016551.2 | NP_057635.1 | 5, 6 | 936 | ||
| XM_005253391.3 | XP_005253448.1 | 5, 6 | 957 | |||
| XM_011520690.2 | XP_011518992.1 | 4, 5 | 799 | |||
| XM_017019463.1 | XP_016874952.1 | 5, 6 | 1047 | |||
| XM_017019464.1 | XP_016874953.1 | 3, 4 | 609 | |||
| XM_017019465.1 | XP_016874954.1 | 3, 4 | 609 | |||
| XM_017019466.1 | XP_016874955.1 | 4, 5 | 776 | |||
| GenBank mRNA | AB032470.1 | - | 5, 6 | 735 | ||
| AK002031.1 | - | 5, 6 | 702 | |||
| AK023085.1 | - | 5, 6 | 924 | |||
| BC005176.2 | - | 5, 6 | 861 | 3505386 | ||

TM7SF3
Genomic Map