siRNA Details
| Gene Symbol: UBQLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017481.2 | NP_059509.1 | 2 | 235 | ||
| XM_011520144.2 | XP_011518446.1 | 2 | 336 | |||
| XM_011520145.2 | XP_011518447.1 | 2 | 305 | |||
| GenBank mRNA | AF230481.1 | - | 2 | 242 | ||
| BC036743.1 | - | 2 | 250 | 5167911 | ||

UBQLN3
Genomic Map