siRNA Details
| Gene Symbol: RBBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198719.1 | NP_001185648.1 | 3 | 523 | ||
| NM_002893.3 | NP_002884.1 | 3 | 535 | |||
| GenBank mRNA | AK091911.1 | - | 3 | 520 | ||
| AK291567.1 | - | 3 | 279 | |||
| BC114500.1 | - | 3 | 433 | 40083852 | ||
| BC114501.1 | - | 3 | 433 | 40083851 | ||
| CR541701.1 | - | 3 | 177 | |||
| DA924163.1 | - | 3 | 523 | |||
| U35143.1 | - | 3 | 438 | |||
| X72841.1 | - | 3 | 462 | |||

RBBP7
Genomic Map