siRNA Details
| Gene Symbol: EML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145076.2 | NP_001138548.1 | 3, 4 | 620 | ||
| NM_019063.4 | NP_061936.2 | 4, 5 | 794 | |||
| XM_005264267.1 | XP_005264324.1 | 4, 5 | 774 | |||
| XM_005264268.3 | XP_005264325.1 | 4, 5 | 774 | |||
| XM_006711991.1 | XP_006712054.1 | 3, 4 | 600 | |||
| XM_006711992.2 | XP_006712055.1 | 5, 6 | 895 | |||
| GenBank mRNA | AF177377.2 | - | 4, 5 | 756 | ||
| AK314702.1 | - | 4, 5 | 768 | |||
| BC140845.1 | - | 4, 5 | 717 | 9021713 | ||
| BC146799.1 | - | 3, 4 | 594 | 8860304 | ||

EML4
Genomic Map