siRNA Details
| Gene Symbol: ENTPD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020354.3 | NP_065087.1 | 3 | 358 | ||
| GenBank mRNA | AF269255.1 | - | 3 | 323 | ||
| AK055540.1 | - | 3 | 358 | |||
| AK309735.1 | - | 4 | 541 | |||
| AK314542.1 | - | 3 | 416 | |||
| BC119008.1 | - | 3 | 324 | 40085532 | ||
| BC122857.1 | - | 3 | 324 | 40085531 | ||

ENTPD7
Genomic Map