siRNA Details
| Gene Symbol: ANGPTL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021146.3 | NP_066969.1 | 1 | 383 | ||
| XM_017000004.1 | XP_016855493.1 | 1 | 348 | |||
| GenBank mRNA | AB593026.1 | - | 1 | 317 | ||
| AB593027.1 | - | 1 | 384 | |||
| AK313716.1 | - | 1 | 349 | |||
| AY358301.1 | - | 1 | 306 | |||
| BC001881.1 | - | 1 | 331 | 3544149 | ||
| BT009802.1 | - | 1 | 92 | |||
| CA391009.1 | - | 1 | 383 | |||
| Y16132.1 | - | 1 | 99 | |||

ANGPTL7
Genomic Map